Safety glasses, Virtua™
Proveedor: 3M
71500-00002 715001AF 715001AF-BULK VIRTUA0
MMMA71500-00002EA 7.4 EUR
MMMA71500-00002 MMMA715001AF MMMA715001AF-BULK MMMAVIRTUA0
Safety glasses, Virtua™
Gafas
Virtua™ safety glasses feature a rimless, wrap-around design for full coverage, and contoured flexible temples to ensure comfort. The integrated side-shields provide added protection from flying objects.
  • Economical basic unisex safety glasses for single-use protection
  • Wrap-around design improves the security of fit and prevents slipping
  • Design provides maximum clarity, excellent coverage, and a wide field of vision
  • Easy to clean
Get affordable, single-use eye protection for on-site visitors with Virtua™ safety glasses. These safety glasses feature a wrap-around design that ensures full coverage and a wide field of view. The contoured flexible temple arms ensure a close and comfortable fit, and the lens provides maximum clarity and features an anti-scratch coating for increased durability.