2-(4-Hidroxifenil)etanol ≥98%

Proveedor: Cayman Chemical
Atención

27600-100 27600-250 27600-500
CAYM27600-100EA 137 EUR
CAYM27600-100 CAYM27600-250 CAYM27600-500
2-(4-Hidroxifenil)etanol ≥98%
2-(4-Hidroxifenil)etanol

Tyrosol is a phenol that has been found in olives and olive oil and has diverse biological activities.


Precaución: This product is not for human or veterinary use.

Formula: C₈H₁₀O₂
Temperatura de almacenaje: Congelador
Número MDL: MFCD00002902
Núm. CAS: 501-94-0

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR