β-Methasone valerate

Proveedor: Thermo Fisher Scientific
Peligro

464930010 464930050
ACRO464930010EA 126 EUR
ACRO464930010 ACRO464930050
β-Methasone valerate
beta-Methasone valerate
Formula: C₂₇H₃₇FO₆
MW: 476.58 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00867446
Núm. CAS: 2152-44-5
EINECS: 218-439-3

Order Now

Resultados test especificaciones

Appearance White to almost white powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Loss on drying ≤ 0.5% (1g,105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR