Methyl Salicylate, Reference Standards

Proveedor: USP

1437450 1437461
1437450.EA 364 EUR
1437450. 1437461.
Methyl Salicylate, Reference Standards
Normativas Estándares orgánicos

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certificaciones: US Pharmacopea Reference Standard

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

ESPECIFICACIONES

Descripción Methyl Salicylate Methyl Salicylate Related Compound A
Synonym(s) Dimethyl 4-hydroxyisophthalate
Environmentally Preferable
CAS No. 119-36-8 5985-24-0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR