Trimipramine Related Compounds, Reference Standard

Proveedor: USP

1692709 1692710
1692709.EA 282 EUR
1692709. 1692710.
Trimipramine Related Compounds, Reference Standard
Normativas Estándares orgánicos

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certificaciones: US Pharmacopea Reference Standard

Order Now

ESPECIFICACIONES

Descripción Trimipramine Maleate Trimipramine Related Compound A
Synonym(s) dehydro trimipramine
Environmentally Preferable
CAS No. 521-78-8 315-69-5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR