High-capacity micro balances, Cubis® MSA
Proveedor: Sartorius Balances
MSA36P-2S00-DH MSA66P-2S00-DH
611-5269EA 0 EUR
611-5269 611-5270
High-capacity micro balances, Cubis® MSA
Balanzas
Cubis® MSA high-capacity micro balances offer readability of 2,1 g and a maximum weighing capacity up to 70200 g.
  • Modular design offering the widest variety of customisable versions
  • Settings configurable to user-specific requirements
  • Customised integration into existing applications by Q-Apps
  • Motorised levelling function
  • The highest accuracy, even for the smallest sample quantities
The Cubis® modular system, consisting of display and control units, weighing modules, draft shields and interface, modules and an extensive range of accessories, enables the balance to be customized to any weighing tasks. The metrological specifications and equipment features of Cubis® set it apart from other weighing instruments far beyond the usual standards of premium laboratory balances. With a finely graduated range of weighing capacities of up to 70 kg and readabilities from 0.1 μg to 1 g, Cubis® offers the ideal choice of model for any application.