GOJO® Mild Foam Hand Wash Bottle
Proveedor: GOJO
GOJO®
115-3475EA 81.9 EUR
115-3475
GOJO® Mild Foam Hand Wash Bottle
Limpieza de la piel Jabones y productos para la piel
A foam hand wash that is gentle to the skin and is both unfragranced and colourless.
  • Suitable for sensitive skin and formulated for high frequency use
  • Broad Spectrum efficacy (bacteria, yeasts, and virus)
  • Passes EN 1499 in 30 seconds
  • Targets food specific pathogens such as Salmonella, Listeria, and E.coli
  • Unfragranced and colourless
  • Halal certified
Mild enough for high-frequency washing, this unfragranced and colourless hand soap is enriched with a moisturising plant-based agent.
Certificaciones: Food taint test compliant: BS EN ISO 4120:2007.