Benzoato de denatonio, anhidro ≥99.5%, granular NF

Proveedor: Spectrum Chemical

DE101-25GM DE101-5GM DE101-1KGBL
SPCMDE101-25GMEA 1380 EUR
SPCMDE101-25GM SPCMDE101-5GM SPCMDE101-1KGBL
Benzoato de denatonio, anhidro ≥99.5%, granular NF
Benzoato de denatonio

Denatonium Benzoate, Anhydrous, Granular, NF is used as a denaturant and masking agent.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formula: C₂₈H₃₄N₂O₃
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00031578
Núm. CAS: 3734-33-6
EINECS: 223-095-2
ADR: 5.2,II

Order Now

Resultados test especificaciones

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR