Food processing gloves, AlphaTec® 37-320
Proveedor: Ansell
37-320/10 37-320/7 37-320/8 37-320/11 37-320/9
ANSE37-320/10EA 16.1 EUR
ANSE37-320/10 ANSE37-320/7 ANSE37-320/8 ANSE37-320/11 ANSE37-320/9
Food processing gloves, AlphaTec® 37-320
Guantes
These unflocked nitrile rubber gloves are ideal for various food processing applications.
  • 100% latex-free: For Type 1 allergy protection
  • Unflocked to prevent the risk of food contamination
  • NBR coating for lightweight chemical protection
  • Long cuff extends to the forearm
  • May be used on their own or as an overglove
The 8 mil/0,2 mm thickness allows dexterity and tactility without a loss of durability. NBR offering protection against light acids, greases, oils, and liquids.
Recommended for: Dairy processing, fruit and vegetable processing, poultryMore Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable
Latex/No contiene Latex No contiene Latex
Material Nitrile
Acabado NBR coating
Tipo de puño Puño tipo guante
Espesor de la palma 0,2 mm
Normativa EN EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (2001X)
Sterility Non sterile
Antiestático +
Aprobados para contacto con alimentos +