Cleaning containers, stainless steel
Proveedor: asecos
8570 8633
MSGA8570EA 308 EUR
MSGA8570 MSGA8633
Cleaning containers, stainless steel
Depósitos Depósitos cilíndricos
Cleaning and immersion containers made from stainless steel.
  • High resistance even to aggressive cleaning fluids
  • Clamps keep the lid closed
When closing the lid the immersion sieve is pressed down, pushing the parts to be cleaned into the cleaning fluids.
Información para pedidos: Small parts baskets are available separately.