PolarPack® Foam Brick, Ice Pack Refrigerant Ice Bricks
Proveedor: Sonoco
PolarPack®
SCATFPP7BEA 0 EUR
SCATFPP18 SCATFPP17 SCATFPP7B SCATFPP31 SCATFPP64 SCATFPP15 SCATFPP56 SCATFPP24
PolarPack® Foam Brick, Ice Pack Refrigerant Ice Bricks
Envases fríos
PolarPack® gel brick coolant pack refrigerants provide thermal solutions for temperature critical packages during transit and are an alternative to wet ice packs.
  • Broad selection available
  • Reliable temperature protection
  • Never change shape during freezing
Available in a broad selection of styles and a multitude of sizes, they’re designed to provide reliable temperature protection for a vast array of products and packages.
These semi-rigid foam bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.