Sodium propionate hydrate ≥99.0% NF
Proveedor: Spectrum Chemical
|
Atención
|
S1418-12KG
S1418-2.5KG
S1418-45KGBL
SPCMS1418-12KGEA
1030
EUR
SPCMS1418-12KG
SPCMS1418-2.5KG
SPCMS1418-45KGBL
Sodium propionate hydrate ≥99.0% NF
Sodium propionate hydrate
Sodium Propionate, NF is a preservative excipient.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₃H₅NaO₂·xH₂O Temperatura de almacenaje: Ambiente |
Núm. CAS:
6700-17-0 |
Resultados test especificaciones
Assay (Dried Basis) | 99.0 - 100.5% |
Alkalinity | To Pass Test |
Water Determination | max. 1.0 % |
Heavy Metals | max. 10 ppm |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...