Quanti-Cult Plus™ Escherichia coli ATCC™ 8739™
Proveedor: THERMO FISHER DIAGNOSTICS
R4717085 R4737085
OXOIR4717085EA 1446 EUR
OXOIR4717085 OXOIR4737085
Quanti-Cult Plus™ Escherichia coli ATCC™ 8739™
Tests para microbiología Tests de microorganismos
Quanti-Cult Plus™ Escherichia coli ATCC™ 8739™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers less than 100 colony forming units (CFUs) in a single 0,1 ml inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Envase: Includes 10 preserved organism vials and 10 rehydration vials.