Quanti-Cult Plus™ Salmonella enterica serovar Typhimurium ATCC™ 13311™
Proveedor: Thermo Fisher Scientific
Quanti-Cult Plus™
OXOIR4716100EA 1446 EUR
OXOIR4716100
Quanti-Cult Plus™ Salmonella enterica serovar Typhimurium ATCC™ 13311™
Tests para microbiología Tests de microorganismos
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 13311™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Envase: Includes 10 preserved organism vials and 10 rehydration vials.