Quanti-Cult Plus™ Streptococcus pyogenes ATCC™ 19615™
Proveedor: Thermo Fisher Scientific
Quanti-Cult Plus™
OXOIR4717000EA 1446 EUR
OXOIR4717000
Quanti-Cult Plus™ Streptococcus pyogenes ATCC™ 19615™
Tests para microbiología Tests de microorganismos
Quanti-Cult Plus™ Streptococcus pyogenes ATCC™ 19615™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
10 preserved organism vials and 10 rehydration vials