ISAspher HPLC Columns, C4
Proveedor: ISERA
A111-E23A00 A111-E23B00 A111-E23C00 A111-E23D00 A111-E23E00 A111-E23F00 A111-E23G00 A111-E23H00 A111-E23J00 A111-E23K00 A111-E23L00 A111-E23M00 A111-E23N00 A111-E23P00 A111-E23Q00 A111-E23R00 A111-E23S00 A111-E23T00 A111-E23U00 A111-E23V00 A111-E23W00 A111-E23X00 A111-E23Y00 A111-E23Z00 A111-E25A00 A111-E25B00 A111-E25C00 A111-E25D00 A111-E25E00 A111-E25F00 A111-E25G00 A111-E25H00 A111-E25J00 A111-E25K00 A111-E25L00 A111-E25M00 A111-E25N00 A111-E25P00 A111-E25Q00 A111-E25R00 A111-E25S00 A111-E25T00 A111-E25U00 A111-E25V00 A111-E25W00 A111-E25X00 A111-E25Y00 A111-E25Z00 A111-E29A00 A111-E29B00 A111-E29C00 A111-E29D00 A111-E29E00 A111-E29F00 A111-E29G00 A111-E29H00 A111-E29J00 A111-E29K00 A111-E29L00 A111-E29M00 A111-E29N00 A111-E29P00 A111-E29Q00 A111-E29R00 A111-E29S00 A111-E29T00 A111-E29U00 A111-E29V00 A111-E29W00 A111-E29X00 A111-E29Y00 A111-E29Z00 A111-E75A00 A111-E75B00 A111-E75C00 A111-E75D00 A111-E75E00 A111-E75F00 A111-E75G00 A111-E75H00 A111-E75J00 A111-E75K00 A111-E75L00 A111-E75M00 A111-E75N00 A111-E75P00 A111-E75Q00 A111-E75R00 A111-E75S00 A111-E75T00 A111-E75U00 A111-E75V00 A111-E75W00 A111-E75X00 A111-E75Y00 A111-E75Z00 A113-E25A00 A113-E25B00 A113-E25D00 A113-E25H00 A113-E25J00 A113-E25K00 A113-E25L00 A113-E29A00 A113-E29B00 A113-E29D00 A113-E29H00 A113-E29J00 A113-E29K00 A113-E29L00 A112-E25A30 A112-E25B30 A112-E25C30 A112-E25E30 A112-E25F30 A112-E25G30 A112-E25A00 A112-E25B00 A112-E25C00 A112-E25E00 A112-E25F00 A112-E25G00 A112-E23A30 A112-E23B30 A112-E23C30 A112-E23E30 A112-E23F30 A112-E23G30 A112-E23A00 A112-E23B00 A112-E23C00 A112-E23E00 A112-E23F00 A112-E23G00 A112-E29A30 A112-E29B30 A112-E29C30 A112-E29E30 A112-E29F30 A112-E29G30 A112-E29A00 A112-E29B00 A112-E29C00 A112-E29E00 A112-E29F00 A112-E29G00 A112-E75A30 A112-E75B30 A112-E75C30 A112-E75E30 A112-E75F30 A112-E75G30 A112-E75A00 A112-E75B00 A112-E75C00 A112-E75E00 A112-E75F00 A112-E75G00
ISERA111-E23A00EA 289 EUR
ISERA111-E23A00 ISERA111-E23B00 ISERA111-E23C00 ISERA111-E23D00 ISERA111-E23E00 ISERA111-E23F00 ISERA111-E23G00 ISERA111-E23H00 ISERA111-E23J00 ISERA111-E23K00 ISERA111-E23L00 ISERA111-E23M00 ISERA111-E23N00 ISERA111-E23P00 ISERA111-E23Q00 ISERA111-E23R00 ISERA111-E23S00 ISERA111-E23T00 ISERA111-E23U00 ISERA111-E23V00 ISERA111-E23W00 ISERA111-E23X00 ISERA111-E23Y00 ISERA111-E23Z00 ISERA111-E25A00 ISERA111-E25B00 ISERA111-E25C00 ISERA111-E25D00 ISERA111-E25E00 ISERA111-E25F00 ISERA111-E25G00 ISERA111-E25H00 ISERA111-E25J00 ISERA111-E25K00 ISERA111-E25L00 ISERA111-E25M00 ISERA111-E25N00 ISERA111-E25P00 ISERA111-E25Q00 ISERA111-E25R00 ISERA111-E25S00 ISERA111-E25T00 ISERA111-E25U00 ISERA111-E25V00 ISERA111-E25W00 ISERA111-E25X00 ISERA111-E25Y00 ISERA111-E25Z00 ISERA111-E29A00 ISERA111-E29B00 ISERA111-E29C00 ISERA111-E29D00 ISERA111-E29E00 ISERA111-E29F00 ISERA111-E29G00 ISERA111-E29H00 ISERA111-E29J00 ISERA111-E29K00 ISERA111-E29L00 ISERA111-E29M00 ISERA111-E29N00 ISERA111-E29P00 ISERA111-E29Q00 ISERA111-E29R00 ISERA111-E29S00 ISERA111-E29T00 ISERA111-E29U00 ISERA111-E29V00 ISERA111-E29W00 ISERA111-E29X00 ISERA111-E29Y00 ISERA111-E29Z00 ISERA111-E75A00 ISERA111-E75B00 ISERA111-E75C00 ISERA111-E75D00 ISERA111-E75E00 ISERA111-E75F00 ISERA111-E75G00 ISERA111-E75H00 ISERA111-E75J00 ISERA111-E75K00 ISERA111-E75L00 ISERA111-E75M00 ISERA111-E75N00 ISERA111-E75P00 ISERA111-E75Q00 ISERA111-E75R00 ISERA111-E75S00 ISERA111-E75T00 ISERA111-E75U00 ISERA111-E75V00 ISERA111-E75W00 ISERA111-E75X00 ISERA111-E75Y00 ISERA111-E75Z00 ISERA113-E25A00 ISERA113-E25B00 ISERA113-E25D00 ISERA113-E25H00 ISERA113-E25J00 ISERA113-E25K00 ISERA113-E25L00 ISERA113-E29A00 ISERA113-E29B00 ISERA113-E29D00 ISERA113-E29H00 ISERA113-E29J00 ISERA113-E29K00 ISERA113-E29L00 ISERA112-E25A30 ISERA112-E25B30 ISERA112-E25C30 ISERA112-E25E30 ISERA112-E25F30 ISERA112-E25G30 ISERA112-E25A00 ISERA112-E25B00 ISERA112-E25C00 ISERA112-E25E00 ISERA112-E25F00 ISERA112-E25G00 ISERA112-E23A30 ISERA112-E23B30 ISERA112-E23C30 ISERA112-E23E30 ISERA112-E23F30 ISERA112-E23G30 ISERA112-E23A00 ISERA112-E23B00 ISERA112-E23C00 ISERA112-E23E00 ISERA112-E23F00 ISERA112-E23G00 ISERA112-E29A30 ISERA112-E29B30 ISERA112-E29C30 ISERA112-E29E30 ISERA112-E29F30 ISERA112-E29G30 ISERA112-E29A00 ISERA112-E29B00 ISERA112-E29C00 ISERA112-E29E00 ISERA112-E29F00 ISERA112-E29G00 ISERA112-E75A30 ISERA112-E75B30 ISERA112-E75C30 ISERA112-E75E30 ISERA112-E75F30 ISERA112-E75G30 ISERA112-E75A00 ISERA112-E75B00 ISERA112-E75C00 ISERA112-E75E00 ISERA112-E75F00 ISERA112-E75G00
ISAspher HPLC Columns, C4
Columnas de cromatografía
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µMore Product Information