Chainmail Gloves
Proveedor: Portwest
AC01SIRL AC01SIRM AC01SIRS AC01SIRXL
PRTWAC01SIRLEA 125 EUR
PRTWAC01SIRL PRTWAC01SIRM PRTWAC01SIRS PRTWAC01SIRXL
Chainmail Gloves
Guantes
Perfect for meat processing environments when protection against sharp blades is vital.
  • Made from stainless steel chainmail for maximum protection
  • Ideal for meat processing
  • Ambidextrous design suitable for left- or right-handed users
  • Suited for food processing plants and resistant to greases, animal fats, and oils
  • Can be used in food industries (metal detectable)
  • Sold in singles
Clip fastening for improved hygiene to reduce bacteria build up. Ambidextrous to suit right and left handed users. Sold singly.
Colour: Silver
Certificaciones: EN 1082-1. CE Food Safe. CE certified.