Flame-Resistant Standard Coats
Proveedor: Portwest
FR34NARL FR34NARM FR34NARS FR34NARXL FR34NARXXL FR34NARXXXL
PRTWFR34NARLEA 47.9 EUR
PRTWFR34NARL PRTWFR34NARM PRTWFR34NARS PRTWFR34NARXL PRTWFR34NARXXL PRTWFR34NARXXXL
Flame-Resistant Standard Coats
Ropa de trabajo
Flame-resistant coats are designed to be worn over flame-resistant garments. Constructed with a comfortable, lightweight, highly innovative flame-resistant twill fabric.
  • Three pockets for ample storage
  • Pen division on left pocket
  • Concealed stud front for easy access
  • Lightweight and comfortable
  • 0+ UPF rated fabric to block 98% of UV rays
  • CE-Cat. III
FR standard coats provide protection against radiant, convective, and contact heat. The lightweight design enhances wearer comfort.
Certificaciones: EN ISO 11612 A1+A2, B1, C1, E2, F1. EN ISO 11611 Class 1 A1+A2. EN 1149 -5. ASTM F1959/F1959M-14e1 ATPV 9.1 cal/cm². CE certified.