Human Recombinant CDK2 Protein (His Tag) (from Baculovirus)

Proveedor: Novus Biologicals

NBP2-51887 NBP2-51887-50U
NOVUNBP2-51887EA 1360 EUR
NOVUNBP2-51887 NOVUNBP2-51887-50U
Human Recombinant CDK2 Protein (His Tag) (from Baculovirus)
Proteínas y Péptidos

A recombinant protein with a C-terminal his-tag and corresponding to the amino acids 1 - 298 of human CDK2.

Order Now

ESPECIFICACIONES

Sinónimos de proteínas cell devision kinase 2|cyclin-dependent kinase 2|Cell division protein kinase 2|cdc2-related protein kinase|p33(CDK2)|p33 protein kinase|EC 2.7.11.22|EC 2.7.11
Environmentally Preferable
Tipo de proteína / péptido Recombinant
Especies Human
Fuente Baculovirus
ID del gen CDK2
Número de aminoácidos MENFQKVEKI GEGTYGVVYK ARNKLTGEVV ALKKIRLDTE TEGVPSTAIR EISLLKELNH PNIVKLLDVI HTENKLYLVF EFLHQDLKKF MDASALTGIP LPLIKSYLFQ LLQGLAFCHS HRVLHRDLKP QNLLINTEGA IKLADFGLAR AFGVPVRTYT HEVVTLWYRA PEILLGCKYY STAVDIWSLG CIFAEMVTRR ALFPGDSEID QLFRIFRTLG TPDEVVWPGV TSMPDYKPSF PKWARQDFSK VVPPLDEDGR SLLSQMLHYD PNKRISAKAA LAHPFFQDVT KPVPHLRLLE HHHHHH
Pureza >90% as determined by SDS-PAGE.
Concentración 0.5 mg/ml
Molecular weight 34,9 kDa
Condiciones de almacenamiento Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR