Cerex® WWP2 SPE Columns
Proveedor: Tecan
30147136 30163879 417-0101R
TCAT30147136EA 0 EUR
TCAT30147136 TCAT30163879 TCAT417-0101R
Cerex® WWP2 SPE Columns
Columnas de cromatografía
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
  • Reversed-phase
  • Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.