Cal-520® sodium salt
Proveedor: AAT BIOQUEST
Atención
21135 21136
AATB21135EA 460 EUR
AATB21135 AATB21136
Cal-520® sodium salt
Cal-520® sodium salt
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell impermeant
Precaución: For research use only.
MW: 840.54 g/mol
ADR: 5 II