Trypan 0.1 M, Trypan Purple™ for dead cell labeling
Proveedor: AAT BIOQUEST
Atención
2465 2466
AATB2465EA 307 EUR
AATB2465 AATB2466
Trypan 0.1 M, Trypan Purple™ for dead cell labeling
Trypan
Trypan Purple™ is similar to Trypan Blue in cell permeability.
  • Minimal cytotoxicity
  • High purity
MW: 600 g/mol
ADR: 5 II