Peridinin-Chlorophyll-Protein Complex fluorescent probe
Proveedor: AAT BIOQUEST
Atención
2540 2559
AATB2540EA 585 EUR
AATB2540 AATB2559
Peridinin-Chlorophyll-Protein Complex fluorescent probe
Peridinin-Chlorophyll-Protein Complex
PerCP (Peridinin-chlorophyll-protein complex) is isolated from Dinophyceae sp.
  • High extinction coefficient
  • High quantum efficiency
  • Large Stokes shift
ADR: 5,II