Anti-IgG (H, L) Goat Polyclonal Antibody (iFluor® 405)

Proveedor: AAT BIOQUEST
16444 16731
AATB16444EA 120 EUR
AATB16444 AATB16731
Anti-IgG (H, L) Goat Polyclonal Antibody (iFluor® 405)
Anticuerpos
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • pH-insensitive fluorescence
  • Photostable
  • Improved robustness

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone:
Conjugation: iFluor™ 405
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR