Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 405)

Proveedor: AAT BIOQUEST
16674 16826
AATB16674EA 152 EUR
AATB16674 AATB16826
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 405)
Anticuerpos
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Precaución: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Alexa Fluor® 405
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR