Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 700)

Proveedor: AAT BIOQUEST
16574 16785
AATB16574EA 152 EUR
AATB16574 AATB16785
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 700)
Anticuerpos
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Precaución: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 700
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR