Bucculite™ FdU Cu-Free Cell Proliferation Fluorescence Imaging Kits
Proveedor: AAT BIOQUEST
22305 22315 22320
AATB22305EA 769 EUR
AATB22305 AATB22315 AATB22320
Bucculite™ FdU Cu-Free Cell Proliferation Fluorescence Imaging Kits
Ensayos Ensayos celulares
Monitoring cell proliferation is one of the most reliable methods to assess cell viability, cell cycles and genotoxicity.
  • Environmentally friendly assay
  • Robust protocol
  • Stable signal
  • Optimised for common channels