Alkaline Phosphatase [CIAP]
Proveedor: AAT BIOQUEST
Atención
11900 11901
AATB11900EA 137 EUR
AATB11900 AATB11901
Alkaline Phosphatase [CIAP]
Alkaline Phosphatase [CIAP]
Calf Intestinal Alkaline Phosphatase (CIP or CIAP) nonspecifically catalyzes the dephosphorylation of 5' and 3' ends of DNA and RNA phosphomonoesters.
  • Extensive applications
  • Compatible with either colorimetric or fluorogenic substrates
  • Multiple possible assays
ADR: 5,II