iFluor® 790, Succinimidyl Ester
Proveedor: AAT BIOQUEST
1368 71368 71524 71574
AATB1368EA 307 EUR
AATB1368 AATB71368 AATB71524 AATB71574
iFluor® 790, Succinimidyl Ester
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • Useful For In-Vivo Imaging
Precaución: For research use only.