Rhod-4™ sodium salt

Proveedor: AAT BIOQUEST
Atención

21118 21128
AATB21118EA 769 EUR
AATB21118 AATB21128
Rhod-4™ sodium salt
Rhod-4™ sodium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Long-wavelength emission
  • Cell impermeant


Precaución: For research use only.

MW: 815.64 g/mol
ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR