7-Bromoheptanoic acid 94%
Proveedor: Apollo Scientific
|
Peligro
|
OR62069-25G
OR62069-100G
APOSOR62069-25GEA
40.6
EUR
APOSOR62069-25G
APOSOR62069-100G
7-Bromoheptanoic acid 94%
7-Bromoheptanoic acid
Radiopharmaceutical precursor.
Formula:
C₇H₁₃BrO₂ |
Número MDL:
MFCD00037800 Núm. CAS: 30515-28-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...