Methyl 3-(2,6-dichlorophenyl)acrylate

Proveedor: Apollo Scientific
Atención

OR111147-250MG OR111147-1G
APOSOR111147-250MGEA 168 EUR
APOSOR111147-250MG APOSOR111147-1G
Methyl 3-(2,6-dichlorophenyl)acrylate
Methyl 3-(2,6-dichlorophenyl)acrylate
Formula: C₁₀H₈Cl₂O₂
Número MDL: MFCD08771574
Núm. CAS: 46275-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR