3-Bromo-N-metilanilina 98%
Proveedor: Thermo Scientific
|
Atención
|
387360010
387360050
ACRO387360010EA
52.5
EUR
ACRO387360010
ACRO387360050
3-Bromo-N-metilanilina 98%
3-Bromo-N-metilanilina
Formula:
C₇H₈BrN MW: 186,05 g/mol |
Número MDL:
MFCD05664376 Núm. CAS: 66584-32-5 UN: 2810 |
Resultados test especificaciones
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6110 to 1.6130 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...