2-Amino-5-(trifluorometil)piridina ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A1968-25G A1968-5G
TCIAA1968-25GEA 232 EUR
TCIAA1968-25G TCIAA1968-5G
2-Amino-5-(trifluorometil)piridina ≥97.0% (by GC, titration analysis)
2-Amino-5-(trifluorometil)piridina
Formula: C₆H₅F₃N₂
Número MDL: MFCD00042164
Núm. CAS: 74784-70-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ee120e083ecf14b5f2d48cd2d706558835e05ba&vid=3ee120e083ecf14b5f2d48cd2d706558835e05ba&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR