2-Bromo-4-fluorobenzonitrilo ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
B2744-25G
B2744-5G
TCIAB2744-25GEA
119
EUR
TCIAB2744-25G
TCIAB2744-5G
2-Bromo-4-fluorobenzonitrilo ≥98.0% (por GC)
2-Bromo-4-fluorobenzonitrilo
Formula:
C₇H₃BrFN MW: 200,01 g/mol |
Número MDL:
MFCD00672924 Núm. CAS: 36282-26-5 EINECS: 252-949-7 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...