L-Cysteic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

C0514-1G C0514-25G
TCIAC0514-1GEA 17 EUR
TCIAC0514-1G TCIAC0514-25G
L-Cysteic acid ≥98.0% (by titrimetric analysis)
L-Cysteic acid
Formula: C₃H₇NO₅S
Pto. de fusión: 273 °C
Número MDL: MFCD00007524
Núm. CAS: 498-40-8
EINECS: 207-861-3
Índice Merck: 14,02780
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67ee962b136012ae48dee70fc47b92ba184bbef9&vid=67ee962b136012ae48dee70fc47b92ba184bbef9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +7.6 to +8.4 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR