Cubis® II MCA 226S Ultra-High Resolution Semi-Micro Balances
Proveedor: Sartorius Balances
MCA226S3CE124M MCA226S3S024 MCA226S3S0234I MCA226S3CE23I MCA226S3S0123 MCA226S3S034H MCA226S3S034 MCA226S3CE3H MCA226S3CE34H MCA226S3S034M MCA226S3CE24 MCA226S3S04H MCA226S3S02 MCA226S3CE13H MCA226S3CE234M MCA226S3CE12 MCA226S3CE234H MCA226S3S024M MCA226S3S0M MCA226S3CE1M MCA226S3S0124 MCA226S3S023M MCA226S3S099I MCA226S3S01M MCA226S3CE12I MCA226S3S01I MCA226S3S02M MCA226S3S013H MCA226S3CE23M MCA226S3S02I MCA226S3CE12H MCA226S3CE14M MCA226S3S014 MCA226S3S0123M MCA226S3CE24H MCA226S3CE99M MCA226S3S023I MCA226S3CE4M MCA226S3S024I MCA226S3CE124I MCA226S3S012H MCA226S3CE14 MCA226S3S013I MCA226S3S0124M MCA226S3CE14I MCA226S3CE234I MCA226S3CE123 MCA226S3S034I MCA226S3CE234 MCA226S3CE14H MCA226S3CE34 MCA226S3S023 MCA226S3CE2M MCA226S3CE4I MCA226S3S0123H MCA226S3S0234H MCA226S3S012 MCA226S3S04 MCA226S3CE2 MCA226S3S04I MCA226S3CE4H MCA226S3S0123I MCA226S3S013 MCA226S3CE124H MCA226S3S02H MCA226S3S03 MCA226S3CE1I MCA226S3S012I MCA226S3S04M MCA226S3S012M MCA226S3S03I MCA226S3CE24M MCA226S3S0124I MCA226S3CE4 MCA226S3S0I MCA226S3CE23H MCA226S3S014I MCA226S3CE2I MCA226S3CE3 MCA226S3CE99I MCA226S3CE124 MCA226S3S03H MCA226S3S014M MCA226S3CE2H MCA226S3CE24I MCA226S3CE3I MCA226S3CE123M MCA226S3S03M MCA226S3CE123H MCA226S3CE13 MCA226S3CE13I MCA226S3CE23 MCA226S3CE123I MCA226S3CE34M MCA226S3CE13M MCA226S3S099M MCA226S3CE3M MCA226S3S013M MCA226S3S0234 MCA226S3S014H MCA226S3S023H MCA226S3CE12M MCA226S3S0234M MCA226S3CE34I MCA226S3S024H MCA226S3S0124H MCA226S3CEM MCA226S3CEI
SARBMCA226S3CE124MEA 28922 EUR
SARBMCA226S3CE124M SARBMCA226S3S024 SARBMCA226S3S0234I SARBMCA226S3CE23I SARBMCA226S3S0123 SARBMCA226S3S034H SARBMCA226S3S034 SARBMCA226S3CE3H SARBMCA226S3CE34H SARBMCA226S3S034M SARBMCA226S3CE24 SARBMCA226S3S04H SARBMCA226S3S02 SARBMCA226S3CE13H SARBMCA226S3CE234M SARBMCA226S3CE12 SARBMCA226S3CE234H SARBMCA226S3S024M SARBMCA226S3S0M SARBMCA226S3CE1M SARBMCA226S3S0124 SARBMCA226S3S023M SARBMCA226S3S099I SARBMCA226S3S01M SARBMCA226S3CE12I SARBMCA226S3S01I SARBMCA226S3S02M SARBMCA226S3S013H SARBMCA226S3CE23M SARBMCA226S3S02I SARBMCA226S3CE12H SARBMCA226S3CE14M SARBMCA226S3S014 SARBMCA226S3S0123M SARBMCA226S3CE24H SARBMCA226S3CE99M SARBMCA226S3S023I SARBMCA226S3CE4M SARBMCA226S3S024I SARBMCA226S3CE124I SARBMCA226S3S012H SARBMCA226S3CE14 SARBMCA226S3S013I SARBMCA226S3S0124M SARBMCA226S3CE14I SARBMCA226S3CE234I SARBMCA226S3CE123 SARBMCA226S3S034I SARBMCA226S3CE234 SARBMCA226S3CE14H SARBMCA226S3CE34 SARBMCA226S3S023 SARBMCA226S3CE2M SARBMCA226S3CE4I SARBMCA226S3S0123H SARBMCA226S3S0234H SARBMCA226S3S012 SARBMCA226S3S04 SARBMCA226S3CE2 SARBMCA226S3S04I SARBMCA226S3CE4H SARBMCA226S3S0123I SARBMCA226S3S013 SARBMCA226S3CE124H SARBMCA226S3S02H SARBMCA226S3S03 SARBMCA226S3CE1I SARBMCA226S3S012I SARBMCA226S3S04M SARBMCA226S3S012M SARBMCA226S3S03I SARBMCA226S3CE24M SARBMCA226S3S0124I SARBMCA226S3CE4 SARBMCA226S3S0I SARBMCA226S3CE23H SARBMCA226S3S014I SARBMCA226S3CE2I SARBMCA226S3CE3 SARBMCA226S3CE99I SARBMCA226S3CE124 SARBMCA226S3S03H SARBMCA226S3S014M SARBMCA226S3CE2H SARBMCA226S3CE24I SARBMCA226S3CE3I SARBMCA226S3CE123M SARBMCA226S3S03M SARBMCA226S3CE123H SARBMCA226S3CE13 SARBMCA226S3CE13I SARBMCA226S3CE23 SARBMCA226S3CE123I SARBMCA226S3CE34M SARBMCA226S3CE13M SARBMCA226S3S099M SARBMCA226S3CE3M SARBMCA226S3S013M SARBMCA226S3S0234 SARBMCA226S3S014H SARBMCA226S3S023H SARBMCA226S3CE12M SARBMCA226S3S0234M SARBMCA226S3CE34I SARBMCA226S3S024H SARBMCA226S3S0124H SARBMCA226S3CEM SARBMCA226S3CEI
Cubis® II MCA 226S Ultra-High Resolution Semi-Micro Balances
Balanzas
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCA display - 7"More Product Information