Quanti-Cult™ Pseudomonas aeruginosa ATCC™ 27853™
Proveedor: Thermo Fisher Scientific
Quanti-Cult™
OXOIR4737060EA 102 EUR
OXOIR4737060
Quanti-Cult™ Pseudomonas aeruginosa ATCC™ 27853™
Tests para microbiología Tests de microorganismos
Quanti-Cult™ Pseudomonas aeruginosa ATCC™ 27853™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0,1 ml inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Envase: Includes 10 preserved organism vials and 10 rehydration vials.