Medical Refrigerator, VR
Proveedor: EFFICOLD
VR-300 VR-100 VR-200 VR-400
EFFIVR-300EA 1570 EUR
EFFIVR-300 EFFIVR-100 EFFIVR-200 EFFIVR-400
Medical Refrigerator, VR
Congeladores Congeladores de laboratorio/aplicaciónes generales
Effimed refrigerators for vaccines ensure design, reliability and quality.
  • Operating temperature range 2 to 8 °C
  • Environmentally friendly refrigerant
  • Door with full double-glazing ensuring excellent thermal insulation
  • Automatic defrost system
  • Visual and audible low/high temperature alarm
  • Visual and audible open door alarm
The device is equipped with a validation port for introducing external sensors.
Safe and precise temperature control with a dual digital thermostat system. The second thermostat acts as a safeguard for the main thermostat to ensure proper storage of products. Excellent waterproof interior design with anti-corrosion protection for easy cleaning and disinfection.
The device is equipped with a door lock to prevent access by unauthorised persons.