Laboratory Refrigerator, LR
Proveedor: EFFICOLD
LR-1300 LR-550
EFFILR-1300EA 3830 EUR
EFFILR-1300 EFFILR-550
Laboratory Refrigerator, LR
Congeladores Congeladores de laboratorio/aplicaciónes generales
Safe and precise temperature control with a dual digital thermostat system. The second thermostat acts as a safeguard for the main thermostat to ensure proper storage of products.
  • Operating temperature range 2 to 8 °C
  • Compact design with external panels made of AISI-304 18/10 stainless steel
  • Inner body made of AISI-304 18/10 stainless steel
  • Environmentally friendly coolant
  • Doors with full double-glasing ensuring excellent thermal insulation
  • Refrigerator equipped with 5 shelves
  • Height-adjustable stainless steel legs
  • Hermetic unit with a ventilated condenser
  • Evaporator with anti-corrosion coating
  • Visual and audible low/high temperature alarm
  • Visual and audible open door alarm
  • RS485 port
The device is equipped with a door lock to prevent access by unauthorised persons.