Laboratory Refrigerator, VFV
Proveedor: EFFICOLD
VFV-200 VFV-400
EFFIVFV-200EA 1360 EUR
EFFIVFV-200 EFFIVFV-400
Laboratory Refrigerator, VFV
Congeladores Congeladores de laboratorio/aplicaciónes generales
Environmentally friendly refrigerant is equipped with a door lock to prevent access by unauthorised persons.
  • Excellent waterproof interior design, chamber entirely made of steel
  • Operating temperature −20 °C
  • Solid door
The device is equipped with a validation port for introducing external sensors.