Anti-Rat Rabbit Polyclonal Antibody (CF®583R)

Proveedor: Biotium
20912-500UL 20912-50UL
20912-500ULEA 248 EUR
20912-500UL 20912-50UL
Anti-Rat Rabbit Polyclonal Antibody (CF®583R)
Anticuerpos
Highly cross-adsorbed rabbit anti-rat IgG (H L) secondary antibody labeled with our superior CF® Dyes.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in 6 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • CF® Dye and biotin conjugates: add 0.5 ml distilled water
  • HRP or DNP conjugates: add 1 ml distilled water

This is a highly cross-adsorbed rabbit anti-rat IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimize cross-reactivity, the antibody has been adsorbed against human serum.

Type: Secondary
Antigen: Rat
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR