Carbenicilina, sal disódica 92

Proveedor: ENZO LIFE SCIENCES
Peligro

Sinónimos: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

ALX380270G001 ALX380270M250
ENZOALX380270G001EA 170 EUR
ENZOALX380270G001 ENZOALX380270M250
Carbenicilina, sal disódica 92
Carbenicillin disodium salt

Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-positive and Gram-negative bacteria and Pseudomonas.

Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00077683
Núm. CAS: 4800-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR