U-18666A ≥99%

Proveedor: ENZO LIFE SCIENCES

BMLS2000010 BMLS2000050
ENZOBMLS2000010EA 165 EUR
ENZOBMLS2000010 ENZOBMLS2000050
U-18666A ≥99%
U-18666A

Oxido-squalene cyclase inhibitor


A useful tool for inhibition of cholesterol transport from late endosomes/lysosomes to the endoplasmic reticulum. Inhibits cholesterol transport in a variety of cell types such as macrophages and sympathetic neurons. May be used to induce a Neimann-Pick disease type C (NPC)or NPC-like phenotype in a variety of cell types. Inhibits cholesterol biosynthesis at the level of oxido-squalene cyclase (OSC). Induction of cataracts is a common toxic feature of OSC inhibitors such as U18666A.

Formula: C₂₅H₄₂ClNO₂
Número MDL: MFCD00210908
Núm. CAS: 3039-71-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=652dda2c39f6fe138e1a64fd000c5d3d94a80489&vid=8dee26b26590a41ea44e1efef6f0aa118acb829f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR