TG100115 ≥98% (by HPLC)
Proveedor: ENZO LIFE SCIENCES
Sinónimos:
TG100-115
ENZCHM1390005
ENZCHM1390010
ENZOENZCHM1390005EA
304
EUR
ENZOENZCHM1390005
ENZOENZCHM1390010
TG100115 ≥98% (by HPLC)
TG100115
Inhibitor of PI3Kγ, PI3Kδ, FRAP, Akt and VEGF.
Formula:
C₁₈H₁₄N₆O₂ MW: 346,3 g/mol |
Número MDL:
MFCD16038873 Núm. CAS: 677297-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...