Farnesylthioacetic acid ≥98%
Proveedor: ENZO LIFE SCIENCES
Methyltransferase inhibitor
FTA potently and specifically inhibits methyl esterification of farnesylated proteins such as the γ subunit of transducin (70% at 10µM). It does not act as a substrate for methyl transferase.
Formula:
C₁₇H₂₈O₂S |
Número MDL:
MFCD00216146 Núm. CAS: 135784-48-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...