Glycogen phosphorylase inhibitor ≥99% (by HPLC)
Proveedor: ENZO LIFE SCIENCES
ENZCHM1100001 ENZCHM1100005
ENZOENZCHM1100001EA 127 EUR
ENZOENZCHM1100001 ENZOENZCHM1100005
Glycogen phosphorylase inhibitor ≥99% (by HPLC)
Glycogen phosphorylase inhibitor
Cell-permeable, potent and AMP-competitive inhibitor of glycogen phosphorylase.
Formula: C₁₇H₁₅ClF₂N₄O₄
Núm. CAS: 648926-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba59f0f520fa17e4d03b90328390f4f2ab88b32f&vid=071881a0b92e30ff23411fdfddf087b3e883dbec&prid=web&clid=SAPDAM