TMP-153 ≥98%
Proveedor: ENZO LIFE SCIENCES
BMLEI3170020
BMLEI3170100
ENZOBMLEI3170020EA
84.7
EUR
ENZOBMLEI3170020
ENZOBMLEI3170100
TMP-153 ≥98%
TMP-153
TMP-153 is a potent and selective inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT), IC50=5-10 nM. It inhibits cholesterol absorption and lowers plasma cholesterol in rats and hamsters. ACAT inhibitors improve plasma lipid profile and ameliorate proteinuria in nephrotic animals.
- ACAT inhibitor
White solid.
Soluble in DMSO (>25mg/ml) or ethanol (10mg/ml with warming).
Formula:
C₂₄H₁₈ClF₂N₃O MW: 437.9 g/mol Temperatura de almacenaje: Ambiente |
Núm. CAS:
128831-46-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...