Carnosic acid ≥95% (by HPLC)
Proveedor: ENZO LIFE SCIENCES
PPARgamma activator. Antioxidant.
Naturally occurring phenolic compound with antioxidant properties. Inhibits lipid peroxidation induced by NADH or NADPH oxidation. Peroxisome proliferator-activated receptor γ (PPARγ) activator. Anti-inflammatory. Antimicrobial.
Isolated from Rosmarinus officinalis.
Formula:
C₂₀H₂₈O₄ |
Número MDL:
MFCD02259459 Núm. CAS: 3650-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...