Bicalutamide ≥98% (by HPLC)
Proveedor: ENZO LIFE SCIENCES
Sinónimos:
N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulphonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulphonyl]-2-hydroxy-2-methylpropionamide
ALX270476G001
ALX270476M100
ALX270476M500
ENZOALX270476G001EA
384
EUR
ENZOALX270476G001
ENZOALX270476M100
ENZOALX270476M500
Bicalutamide ≥98% (by HPLC)
Bicalutamide
Androgen receptor inhibitor
Nonsteroidal antiandrogen. Effective in the treatment of prostate cancer. Induces cell death by a pathway that is independent of changes in mitochondrial membrane potential and Bcl-2 action.
Synthetic.
Formula:
C₁₈H₁₄F₄N₂O₄S MW: 430,37 g/mol |
Número MDL:
MFCD00869971 Núm. CAS: 90357-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...